Få mere at vide
2-[4-(Benzyloxy)phenyl]ethanamine-d4, TRC
Artikelnummer. 30341092
Skift visning
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30341092 25 mg
30333460 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30341092 Leverandør Toronto Research Chemicals Leverandørnr. TRCB28897225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[4-(Benzyloxy)phenyl]ethanamine-d4
CAS 110790-10-8
Molekylær formel C15H13D4NO
Mængde 25 mg
Synonym 2-[4-(Benzyloxy)phenyl]ethylamine-d4, 4-Benzyloxyphenethylamine-d4, O-Benzyltyramine-d4, p-Benzyloxyphenylethylamine-d4, 4-(Phenylmethoxy)benzeneethanamine-d4
InChI formel InChI=1S/C15H17NO/c16-11-10-13-6-8-15(9-7-13)17-12-14-4-2-1-3-5-14/h1-9H,10-12,16H2/i10D2,11D2
SMIL [2H]C([2H])(N)C([2H])([2H])c1ccc(OCc2ccccc2)cc1
IUPAC navn 1,1,2,2-tetradeuterio-2-(4-phenylmethoxyphenyl)ethanamine
Molekylvægt (g/mol) 231.33
Formel vægt 231.1561