Få mere at vide
2-(4-Benzhydrylpiperazin-1-yl)ethan-1-ol Dihydrochloride, Mikromol™
Beskrivelse
2-(4-Benzhydrylpiperazin-1-yl)ethan-1-ol Dihydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-(4-Benzhydrylpiperazin-1-yl)ethan-1-ol Dihydrochloride |
| CAS | 108983-83-1 |
| Analyt- eller komponentnavne | 2-(4-Benzhydrylpiperazin-1-yl)ethan-1-ol Dihydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C19 H24 N2 O . 2 Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 4-(Diphenylmethyl)-1-piperazineethanol Hydrochloride (1:2), 1-Piperazineethanol, 4-diphenylmethyl-, dihydrochloride (6CI), 4-Benzhydryl-1-piperazineethanol Dihydrochloride, 2-(4-(Diphenylmethyl)piperazin-1-yl)ethanol Dihydrochloride, Deschlorodesacetic Cetirizine, Cetirizine Related Compound B as Dihydrochloride |
| InChI formel | InChI=1S/C19H24N2O.2ClH/c22-16-15-20-11-13-21(14-12-20)19(17-7-3-1-4-8-17)18-9-5-2-6-10-18;;/h1-10,19,22H,11-16H2;2*1H |
| SMIL | Cl.Cl.OCCN1CCN(CC1)C(c2ccccc2)c3ccccc3 |
| IUPAC navn | 2-(4-benzhydrylpiperazin-1-yl)ethanol;dihydrochloride |
| Vis mere |