missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2-(4-Aminophenyl)-4-(4-pyridyl)thiazole, 97%
Brand: Alfa Aesar™ H52250.03
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
1153000-38-4 | |
253.323 | |
BNYZGNWCDWUMSU-UHFFFAOYSA-N | |
43667817 | |
C1=CC(=CC=C1C2=NC(=CS2)C3=CC=NC=C3)N |
C14H11N3S | |
MFCD12114461 | |
2-4-aminophenyl-4-4-pyridyl thiazole, 4-4-pyridin-4-yl-1,3-thiazol-2-yl aniline | |
4-(4-pyridin-4-yl-1,3-thiazol-2-yl)aniline |
Specifications
2-(4-Aminophenyl)-4-(4-pyridyl)thiazole | |
1g | |
C14H11N3S | |
2-4-aminophenyl-4-4-pyridyl thiazole, 4-4-pyridin-4-yl-1,3-thiazol-2-yl aniline | |
C1=CC(=CC=C1C2=NC(=CS2)C3=CC=NC=C3)N | |
253.323 | |
253.32 |
Air Sensitive | |
1153000-38-4 | |
MFCD12114461 | |
BNYZGNWCDWUMSU-UHFFFAOYSA-N | |
4-(4-pyridin-4-yl-1,3-thiazol-2-yl)aniline | |
43667817 | |
97% |