Få mere at vide
2,4,7,9-Tetramethyl-5-decyne-4,7-diol, TRC
Artikelnummer. 30137467
Skift visning
Mængde:
5 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137467 100 g
30136452 50 g
30012739 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137467 Leverandør Toronto Research Chemicals Leverandørnr. TRCT302500100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 126-86-3
Molekylær formel C14 H26 O2
Molekylvægt (g/mol) 226.36
Synonym 2,4,7,9-Tetramethyl-5-decyne-4,7-diol, 1,4-Diisobutyl-1,4-dimethylbutynediol, 2,4,7,9-Tetramethyl-5-decyn-4,7-diol, Acetylenol E 00, NSC 5630, Olfine AK 02, Olfine PD 301, Olfine STG-E, PD 30, Surfynol 104, Surfynol 104A, Surfynol 104BC, Surfynol 104E, Surfynol 104H, Surfynol 104PA, Surfynol CT 111, Surfynol PG 50, Surfynol TG, Zetasperse 1200;
IUPAC navn 2,3,8,9-tetramethyldec-5-yne-3,8-diol
SMIL CC(C)C(C)(O)CC#CCC(C)(O)C(C)C
Tekniske data
Kemisk navn eller materiale 2,4,7,9-Tetramethyl-5-decyne-4,7-diol (mix of (±) and meso)
Mængde 100 g
InChI formel InChI=1S/C14H26O2/c1-11(2)13(5,15)9-7-8-10-14(6,16)12(3)4/h11-12,15-16H,9-10H2,1-6H3
Formel vægt 226.1933