Learn More
2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine, Mass Spec Std, Thermo Scientific™ Product Code.: 11399315
2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine, Mass Spec Std, Thermo Scientific™
Product Code. p-7058458
100 mg, 100mg
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 11399315
Brand: Thermo Scientific Alfa Aesar L16678.MC
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine is used as a mass spectrometry standard.
Solubility
Insoluble in water.
Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21674-38-4
Molecular Formula C24F45N3
Molecular Weight (g/mol) 1185.213
MDL Number MFCD00042439
InChI Key BFAUAMWYFFXWLX-UHFFFAOYSA-N
Synonym 2,4,6-tris pentadecafluoroheptyl-1,3,5-triazine, 2,4,6-tris perfluoroheptyl-1,3,5-triazine, 2,4,6-tris 1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptyl-1,3,5-triazine, tris 1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptyl-1,3,5-triazine, acmc-20aoej, perfluorotriheptyl-s-triazine, tri perfluoroheptyl-s-triazine, tris perfluoroheptyl-s-triazine, tris pentadecafluoroheptyl-1,3,5-triazine, 1,3,5-triazine, 2,4,6-tris pentadecafluoroheptyl
PubChem CID 550641
IUPAC Name 2,4,6-tris(1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptyl)-1,3,5-triazine
SMILES C1(=NC(=NC(=N1)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
Specifications
Melting Point 29°C to 31°C
Boiling Point 150°C (1.5 mmHg)
Quantity 100 mg
Solubility Information Insoluble in water.
Formula Weight 1185.22
Chemical Name or Material 2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine, Mass Spec Standard
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberLF005211 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot Number10139067 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberB07X020 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberF21R026 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberK04X012 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction
Product Title
2,4,6-Tris(perfluoroheptyl)-1,3,5-triazine, Mass Spec Std, Thermo Scientific™ >