Få mere at vide
2,4,6-Trimethyl-5-nitrobenzene-d11, TRC
Artikelnummer. 30451154
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30451154 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451154 Leverandør Toronto Research Chemicals Leverandørnr. TRCT79646210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4,6-Trimethyl-5-nitrobenzene-d11
CAS 1126138-12-2
Molekylær formel C9D11NO2
Mængde 10 mg
Synonym 2,4,6-Tri(methyl-d3)-5-nitro-benzene-1,3-d2
InChI formel InChI=1S/C9H11NO2/c1-6-4-7(2)9(10(11)12)8(3)5-6/h4-5H,1-3H3/i1D3,2D3,3D3,4D,5D
SMIL [2H]c1c(c([2H])c(c(c1C([2H])([2H])[2H])[N+](=O)[O-])C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1,3-dideuterio-5-nitro-2,4,6-tris(trideuteriomethyl)benzene
Molekylvægt (g/mol) 176.26
Formel vægt 176.148