missing translation for 'onlineSavingsMsg'
Learn More
2,4,6-Triisopropylbenzenesulfonyl hydrazide Product Code.: 11343407

2,4,6-Triisopropylbenzenesulfonyl hydrazide

Product Code. p-7049673
0.5 g, 0.50g
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.50g
2g
This item is not returnable. View return policy

Product Code. 11343407

Brand: Thermo Scientific Alfa Aesar 017976.02

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

As a reagent for generation of diimide | CAS: 39085-59-1 | C15H26N2O2S | 298.45 g/mol

As a reagent for generation of diimide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39085-59-1
Molecular Formula C15H26N2O2S
Molecular Weight (g/mol) 298.45
MDL Number MFCD00014750
InChI Key UGRVYFQFDZRNMQ-UHFFFAOYSA-N
Synonym 2,4,6-triisopropylbenzenesulfonohydrazide, 2,4,6-triisopropylbenzenesulfonyl hydrazide, 2,4,6-triisopropylbenzenesulphonohydrazide, 2,4,6-tri propan-2-yl benzenesulfonohydrazide, benzenesulfonic acid, 2,4,6-tris 1-methylethyl-, hydrazide, tpsh, ksc918s7f, 2,4,6-triisopropylbenzene sulfonohydrazide, 2,4,6-triisopropylbenzenesulfono hydrazide, 2,4,6-triisopropylbenzene sulphonohydrazideShow MoreShow Less
PubChem CID 359333
IUPAC Name 2,4,6-tri(propan-2-yl)benzenesulfonohydrazide
SMILES CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(=O)(=O)NN

Specifications

Melting Point 110°C to 112°C (decomposition)
Quantity 0.5 g
Beilstein 2145001
Sensitivity Moisture sensitive
Formula Weight 298.58
Physical Form Crystalline
Chemical Name or Material 2, 4, 6-Triisopropylbenzenesulfonyl hydrazide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberZ10K035 Certificate TypeCertificate of Analysis Date11/12/2024 Product Code017976.02,017976.04
Lot NumberU01J027 Certificate TypeCertificate of Analysis Date08/08/2023 Product Code017976.02,017976.04
Lot NumberS29J002 Certificate TypeCertificate of Analysis Date14/07/2023 Product Code017976,017976.02,017976.04
Lot NumberS13J016 Certificate TypeCertificate of Analysis Date21/06/2023 Product Code017976,017976.02,017976.04
Lot NumberF28R020 Certificate TypeCertificate of Analysis Date07/12/2021 Product Code017976.02,017976.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4,6-Triisopropylbenzenesulfonyl hydrazide >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.