Få mere at vide
2,4,6-Trihydroxybenzoic Acid, TRC
Artikelnummer. 30456682
Skift visning
Mængde:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30456682 10 g
30454383 100 g
30449586 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30456682 Leverandør Toronto Research Chemicals Leverandørnr. TRCT79506510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 83-30-7
Molekylær formel C7 H6 O5
Molekylvægt (g/mol) 170.12
Synonym 2,4,6-Trihydroxybenzenecarboxylic Acid, NSC 36720, Phloroglucinic Acid, Phloroglucinolcarboxylic Acid
IUPAC navn 2,4,6-trihydroxybenzoic acid
SMIL OC(=O)c1c(O)cc(O)cc1O
Tekniske data
Kemisk navn eller materiale Phloroglucinol carboxylic acid
Mængde 10 g
InChI formel InChI=1S/C7H6O5/c8-3-1-4(9)6(7(11)12)5(10)2-3/h1-2,8-10H,(H,11,12)
Formel vægt 170.0215