missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorophenol, 99%, Thermo Scientific™

Catalog Number 11387666
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2268-17-9
C6H3F3O
148.084
MFCD00061216
QQFWMPUXPLBWTG-UHFFFAOYSA-N
519974
2,4,6-trifluorophenol
C1=C(C=C(C(=C1F)O)F)F
This item is not returnable. View return policy
2268-17-9
C6H3F3O
148.084
MFCD00061216
QQFWMPUXPLBWTG-UHFFFAOYSA-N
519974
2,4,6-trifluorophenol
C1=C(C=C(C(=C1F)O)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 47°C to 52°C
Quantity 1 g
UN Number UN3261
Beilstein 2089551
Synonym phenol, 2,4,6-trifluoro, 2,4,6-trifluoro-phenol, pubchem2317, acmc-1cod0, phenol,2,4,6-trifluoro, 2,4,6-trifluorophenol
Formula Weight 148.09
Percent Purity 99%
Chemical Name or Material 2,4,6-Trifluorophenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'