missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trifluorobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7031612
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
220239-64-5
C6H2ClF3O2S
230.585
MFCD01091015
XINCBNCLCIQIJM-UHFFFAOYSA-N
2,4,6-trifluorobenzene-1-sulfonyl chloride, 2,4,6-trifluorobenzenesulphonyl chloride, pubchem5181, chloro 2,4,6-trifluorophenyl sulfone, 2,4,6-trifluorobenzenesulfonylchloride, 2,4,6-trifluorophenylsulfonyl chloride, benzenesulfonylchloride, 2,4,6-trifluoro, 2,4,6-trifluorobenzenesulfonyl chloride, benzenesulfonyl chloride, 2,4,6-trifluoro-9ci
2776983
2,4,6-trifluorobenzenesulfonyl chloride
C1=C(C=C(C(=C1F)S(=O)(=O)Cl)F)F
This item is not returnable. View return policy
220239-64-5
C6H2ClF3O2S
230.585
MFCD01091015
XINCBNCLCIQIJM-UHFFFAOYSA-N
2,4,6-trifluorobenzene-1-sulfonyl chloride, 2,4,6-trifluorobenzenesulphonyl chloride, pubchem5181, chloro 2,4,6-trifluorophenyl sulfone, 2,4,6-trifluorobenzenesulfonylchloride, 2,4,6-trifluorophenylsulfonyl chloride, benzenesulfonylchloride, 2,4,6-trifluoro, 2,4,6-trifluorobenzenesulfonyl chloride, benzenesulfonyl chloride, 2,4,6-trifluoro-9ci
2776983
2,4,6-trifluorobenzenesulfonyl chloride
C1=C(C=C(C(=C1F)S(=O)(=O)Cl)F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 230.59
Percent Purity 97%
Chemical Name or Material 2,4,6-Trifluorobenzenesulfonyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'