Få mere at vide
2,4,6-Trifluorobenzenemethanamine-d4,15N Deuterium Chloride, TRC
Artikelnummer. 30418432
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30418432 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418432 Leverandør Toronto Research Chemicals Leverandørnr. TRCT184652100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4,6-Trifluorobenzenemethanamine-d4,15N Deuterium Chloride
CAS 2732917-43-8
Molekylær formel C7H2D4F315N • DCl
Mængde 100 mg
InChI formel InChI=1S/C7H6F3N.ClH/c8-4-1-6(9)5(3-11)7(10)2-4;/h1-2H,3,11H2;1H/i3D2,11+1;/hD3
SMIL FC1=CC(F)=CC(F)=C1C([2H])([2H])[15N]([2H])[2H].[2H]Cl
IUPAC navn (2,4,6-trifluorophenyl)methan-d2-amine-d2-15N deuterium chloride
Molekylvægt (g/mol) 203.61
Formel vægt 203.05