Få mere at vide
2,4,6-Trifluorobenzene-1,3,5-tricarbonitrile, TRC
Artikelnummer. 30455008
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30455008 10 mg
30455751 50 mg
30451150 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30455008 Leverandør Toronto Research Chemicals Leverandørnr. TRCT89991010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3638-97-9
Molekylær formel C9N3F3
Molekylvægt (g/mol) 207.11
IUPAC navn 2,4,6-trifluorobenzene-1,3,5-tricarbonitrile
SMIL Fc1c(C#N)c(F)c(C#N)c(F)c1C#N
Tekniske data
Kemisk navn eller materiale 2,4,6-Trifluorobenzene-1,3,5-tricarbonitrile
Mængde 10 mg
InChI formel InChI=1S/C9F3N3/c10-7-4(1-13)8(11)6(3-15)9(12)5(7)2-14
Formel vægt 207.0044