missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trichloropyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-4089824
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
16063-69-7
C5H2Cl3N
182.44
MFCD00971928
FJNNGKMAGDPVIU-UHFFFAOYSA-N
pyridine, 2,4,6-trichloro, 2,4,6-trichloro-pyridine, pubchem11020, acmc-1bu24, ksc497g8t, 2,4,6-tris chloranyl pyridine, 2,4,6-trichloropyridine
27665
2,4,6-trichloropyridine
C1=C(C=C(N=C1Cl)Cl)Cl
This item is not returnable. View return policy
16063-69-7
C5H2Cl3N
182.44
MFCD00971928
FJNNGKMAGDPVIU-UHFFFAOYSA-N
pyridine, 2,4,6-trichloro, 2,4,6-trichloro-pyridine, pubchem11020, acmc-1bu24, ksc497g8t, 2,4,6-tris chloranyl pyridine, 2,4,6-trichloropyridine
27665
2,4,6-trichloropyridine
C1=C(C=C(N=C1Cl)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 33°C to 37°C
Color Brown-Yellow to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 182.44
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4, 6-Trichloropyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4,6-Trichloropyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'