Få mere at vide
2,4,6-Trichlorophen-3,5-d2-ol, TRC
Artikelnummer. 30452522
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30452522 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452522 Leverandør Toronto Research Chemicals Leverandørnr. TRCT77415310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4,6-Trichlorophenol ring-d2
CAS 93951-80-5
Molekylær formel C6 2H2 H Cl3 O
Mængde 10 mg
Synonym Phen-3,5-d2-ol','2,4,6-trichloro-, 2,4,6-Trichlorophen-3,5-d2-ol, 2,4,6-Trichlorophenol D2, 2,4,6-Trichlorophenol (ring-D2)
InChI formel InChI=1S/C6H3Cl3O/c7-3-1-4(8)6(10)5(9)2-3/h1-2,10H/i1D,2D
SMIL [2H]c1c(Cl)c([2H])c(Cl)c(O)c1Cl
IUPAC navn 2,4,6-trichloro-3,5-dideuteriophenol
Molekylvægt (g/mol) 199.46
Formel vægt 197.9375