Få mere at vide
2,4,6-Trichlorobiphenyl-2',3',4',5',6'-d5, CDN
Artikelnummer. 30325076
Skift visning
Mængde:
0.01 g
Artikelnummer. Mængde
30325076 0.01 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325076 Leverandør CDN Leverandørnr. CDND69380.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4,6-Trichlorobiphenyl-2',3',4',5',6'-D5
CAS 1219794-85-0
Molekylær formel C12 D5 H2 Cl3
Mængde 0.01 g
Synonym 2,4,6-Trichlorobiphenyl-2',3',4',5',6'-d5, 2,4,6-Trichloro-1,1'-biphenyl-2',3',4',5',6'-d5, 2,4,6-Trichlorobiphenyl-d5, PCB 30-d5, 2,4,6-TriCB-2',3',4',5',6'-d5
InChI formel InChI=1 S/C12H7Cl3/c13-9-6-10(14)12(11(15)7-9)8-4-2-1-3-5-8/h1-7 H/i1D,2 D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])c2c(Cl)cc(Cl)cc2Cl
IUPAC navn 1,2,3,4,5-pentadeuterio-6-(2,4,6-trichlorophenyl)benzene
Molekylvægt (g/mol) 262.5738
Formel vægt 260.9927 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity