Få mere at vide
2,4,6-Tribromoanisole-d5, TRC
Artikelnummer. 30456318
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30456318 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30456318 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7718511MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4,6-Tribromoanisole-d5
CAS 1219795-33-1
Molekylær formel C7 D5 Br3 O
Mængde 1 mg
Synonym 2,4,6-Tribromoanisole D5, Benzene-1,3-d2','2,4,6-tribromo-5-(methoxy-d3)- (ACI), 2,4,6-Tribromo-5-(methoxy-d3)benzene-1,3-d2 (ACI), 2,4,6-Tribromoanisole-d5
InChI formel InChI=1S/C7H5Br3O/c1-11-7-5(9)2-4(8)3-6(7)10/h2-3H,1H3/i1D3,2D,3D
SMIL [2H]c1c(Br)c([2H])c(Br)c(OC([2H])([2H])[2H])c1Br
IUPAC navn 1,3,5-tribromo-2,4-dideuterio-6-(trideuteriomethoxy)benzene
Molekylvægt (g/mol) 349.857
Formel vægt 346.82