Få mere at vide
2,4,6-Tribromoanisole-d5, CDN
Artikelnummer. 30321313
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321313 0.1 g 0.1g
30321036 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321313 Leverandør CDN Leverandørnr. CDND56780.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4,6-Tribromoanisole-d5
CAS 1219795-33-1
Molekylær formel C7 D5 Br3 O
Mængde 0.1 g
Synonym 2,4,6-Tribromoanisole D5, Benzene-1,3-d2, 2,4,6-tribromo-5-(methoxy-d3)- (ACI), 2,4,6-Tribromo-5-(methoxy-d3)benzene-1,3-d2 (ACI), 2,4,6-Tribromoanisole-d5
InChI formel InChI=1 S/C7H5Br3O/c1-11-7-5(9)2-4(8)3-6(7)10/h2-3 H,1H3/i1D3,2 D,3 D
SMIL [2 H]c1c(Br)c([2 H])c(Br)c(OC([2 H])([2 H])[2 H])c1Br
IUPAC navn 1,3,5-tribromo-2,4-dideuterio-6-(trideuteriomethoxy)benzene
Molekylvægt (g/mol) 349.857
Formel vægt 346.82 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity