Få mere at vide
2,4,6-Triamino-5-pyrimidinecarbonitrile, TRC
Artikelnummer. 30006967
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006967 25 mg
30006581 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30006967 Leverandør Toronto Research Chemicals Leverandørnr. TRCT76722025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 465531-97-9
Molekylær formel C5 H6 N6
Molekylvægt (g/mol) 150.14
Synonym 2,4,6-Triamino-5-pyrimidinecarbonitrile, Dicyclanil-descyclopropyl, Descyclopropyldicyclanil
IUPAC navn 2,4,6-triaminopyrimidine-5-carbonitrile
SMIL Nc1nc(N)c(C#N)c(N)n1
Tekniske data
Kemisk navn eller materiale 2,4,6-Triamino-5-pyrimidinecarbonitrile
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C5H6N6/c6-1-2-3(7)10-5(9)11-4(2)8/h(H6,7,8,9,10,11)
Formel vægt 150.0654
Procent renhed >95%