Få mere at vide
2-[4,6-Bis(2,4-dimethylphenyl)-1,3,5-triazin-2-yl]-5-(octyloxy)phenol, TRC
Artikelnummer. 30134307
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134307 10 g
30006511 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134307 Leverandør Toronto Research Chemicals Leverandørnr. TRCB42990010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2725-22-6
Molekylær formel C33 H39 N3 O2
Molekylvægt (g/mol) 509.68
Synonym 2-(4,6-Di-2,4-xylyl-s-triazin-2-yl)-5-(octyloxy)phenol, 2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-n-octyloxyphenyl)-1,3,5-triazine, 2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-octyloxyphenyl)-1,3,5-triazine, 2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-octyloxyphenyl)-s-triazine, 2,6-Bis(2,4-dimethylphenyl)-4-(2-hydroxy-4-octyloxyphenyl)-s-triazine, 2-(4,6-Bis(2,4-dimethylphenyl)-1,3,5-triazine-2-yl)-5-octyloxyphenol, Cyagard UV 1164, Cyasorb 1164, Cyasorb UV 1164, Cytec UV 1164, Tinuvin 1545, UV 1164;
IUPAC navn 2-[4,6-bis(2,4-dimethylphenyl)-1,3,5-triazin-2-yl]-5-octoxyphenol
SMIL CCCCCCCCOc1ccc(c(O)c1)c2nc(nc(n2)c3ccc(C)cc3C)c4ccc(C)cc4C
Tekniske data
Kemisk navn eller materiale 2-[4,6-Bis(2,4-dimethylphenyl)-1,3,5-triazin-2-yl]-5-(octyloxy)phenol
Mængde 10 g
InChI formel InChI=1S/C33H39N3O2/c1-6-7-8-9-10-11-18-38-26-14-17-29(30(37)21-26)33-35-31(27-15-12-22(2)19-24(27)4)34-32(36-33)28-16-13-23(3)20-25(28)5/h12-17,19-21,37H,6-11,18H2,1-5H3
Formel vægt 509.3042