Få mere at vide
2,4,5-Trichlorophenylethanediol, TRC
Artikelnummer. 30456081
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30456081 500 mg
30455731 1 g
30449843 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30456081 Leverandør Toronto Research Chemicals Leverandørnr. TRCT382940500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14299-53-7
Molekylær formel C8 H7 Cl3 O2
Molekylvægt (g/mol) 241.5
Synonym 1-(2,4,5-Trichlorophenyl)-1,2-ethanediol, 2,4,5-Trichlorophenylethanediol
IUPAC navn 1-(2,4,5-trichlorophenyl)ethane-1,2-diol
SMIL OCC(O)c1cc(Cl)c(Cl)cc1Cl
Tekniske data
Kemisk navn eller materiale 2,4,5-Trichlorophenylethanediol
Mængde 500 mg
InChI formel InChI=1S/C8H7Cl3O2/c9-5-2-7(11)6(10)1-4(5)8(13)3-12/h1-2,8,12-13H,3H2
Formel vægt 239.9512