Få mere at vide
2,4,5-Trichlorophenoxyacetic Acid-d4, TRC
Artikelnummer. 30451924
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30451924 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451924 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7741582.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4,5-TD4
CAS 358731-37-0
Molekylær formel C8 2H4 H Cl3 O3
Mængde 2.5 mg
Synonym 2,4,5-T D4, Acetic-d2 acid','(3,4,6-trichlorophenoxy-2,5-d2)- (9CI)
InChI formel InChI=1S/C8H5Cl3O3/c9-4-1-6(11)7(2-5(4)10)14-3-8(12)13/h1-2H,3H2,(H,12,13)/i1D,2D,3D2
SMIL [2H]c1c(Cl)c(Cl)c([2H])c(OC([2H])([2H])C(=O)O)c1Cl
IUPAC navn 2,2-dideuterio-2-(2,4,5-trichloro-3,6-dideuteriophenoxy)acetic acid
Molekylvægt (g/mol) 259.51
Formel vægt 257.9555