Få mere at vide
2,4,5-Trichlorophenoxyacetic Acetic Acid, SPEX CertiPrep™
Artikelnummer. 12979394 Shop alle SPEX Certiprep produkter
Skift visning
Mængde:
1 mL
Pakningsstørrelse:
1ml
Artikelnummer. Mængde unitSize
12979394 1 mL 1ml
1 options
Artikelnummer. 12979394 Leverandør SPEX Certiprep Leverandørnr. S3655
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Indeholder
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
2,4,5-Trichlorophenoxy acetic acid93-76-51000μg/mL
Certificeringer
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Overholdelse
GHS/OSHA Complaint
Tekniske data
Kemisk navn eller materiale 2,4,5-Trichlorophenoxy Acetic Acid
Emballage 2mL amber prescored glass ampule
CAS 93-76-5, 1634-04-4
CAS min % 0.1, 99.9
Koncentration eller sammensætningsnoter Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molekylær formel C8H5Cl3O3
MDL nummer MFCD00004301
Til brug med (udstyr) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI nøgle SMYMJHWAQXWPDB-UHFFFAOYSA-N
SMIL OC(=O)COC1=CC(Cl)=C(Cl)C=C1Cl
IUPAC navn 2-(2,4,5-trichlorophenoxy)acetic acid
Opløsningsmiddel eller Matrix MTBE
Molekylvægt (g/mol) 255.48
PubChem CID 1480
Mængde 1 mL
ChEBI CHEBI:27903
Vis mere Vis mindre
Product intended for laboratory use only