Få mere at vide
(2,4,5-Trichlorophenoxy-3,6-d2)acetic-d2 Acid, CDN
Artikelnummer. 30319124
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319124 0.05 g
30314184 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319124 Leverandør CDN Leverandørnr. CDND55530.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4,5-TD4
CAS 358731-37-0
Molekylær formel C8 2H4 H Cl3 O3
Mængde 0.05 g
InChI formel InChI=1 S/C8H5Cl3O3/c9-4-1-6(11)7(2-5(4)10)14-3-8(12)13/h1-2 H,3H2,(H,12,13)/i1D,2 D,3D2
SMIL [2 H]c1c(Cl)c(Cl)c([2 H])c(OC([2 H])([2 H])C(=O)O)c1Cl
IUPAC navn 2,2-dideuterio-2-(2,4,5-trichloro-3,6-dideuteriophenoxy)acetic acid
Molekylvægt (g/mol) 259.51
Formel vægt 257.9555 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity