Få mere at vide
2,4,5-Trichlorophenol-d2, TRC
Artikelnummer. 30449594
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30449594 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30449594 Leverandør Toronto Research Chemicals Leverandørnr. TRCT77414910MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4,5-Trichlorophenol ring-d2
CAS 93951-82-7
Molekylær formel C6 2H2 H Cl3 O
Mængde 10 mg
Synonym 2,4,5-Trichlorophenol D2 (3,6-D2), Phen-2,5-d2-ol','3,4,6-trichloro- (9CI), 2,4,5-Trichlorophenol (Ring-D2), 2,4,5-Trichlorophenol D2, Phen-2,5-d2-ol','3,4,6-trichloro- (9CI), 2,4,5-Trichlorophenol (Ring-D2), 2,4,5-Trichlorophenol-D2
InChI formel InChI=1S/C6H3Cl3O/c7-3-1-5(9)6(10)2-4(3)8/h1-2,10H/i1D,2D
SMIL [2H]c1c(O)c(Cl)c([2H])c(Cl)c1Cl
IUPAC navn 2,4,5-trichloro-3,6-dideuteriophenol
Molekylvægt (g/mol) 199.46
Formel vægt 197.9375