Få mere at vide
2,4,5-Trichlorophenol-3,6-d2, CDN
Artikelnummer. 30322237
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322237 0.25 g -
30313681 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322237 Leverandør CDN Leverandørnr. CDND29350.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,4,5-Trichlorophenol ring-d2
CAS 93951-82-7
Molekylær formel C6 2H2 H Cl3 O
Mængde 0.25 g
Synonym Phen-2,5-d2-ol, 3,4,6-trichloro- (9 CI), 2,4,5-Trichlorophenol (Ring-D2), 2,4,5-Trichlorophenol-D2
InChI formel InChI=1 S/C6H3Cl3O/c7-3-1-5(9)6(10)2-4(3)8/h1-2,10 H/i1D,2 D
SMIL [2 H]c1c(O)c(Cl)c([2 H])c(Cl)c1Cl
IUPAC navn 2,4,5-trichloro-3,6-dideuteriophenol
Molekylvægt (g/mol) 199.46
Formel vægt 197.9375 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity