missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trichlorobenzoic Acid, TRC

Product Code. 30348160
Change view
Click to view available options
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30348160 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348160 Supplier Toronto Research Chemicals Supplier No. TRCB58562350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2,4,5-Trichlorobenzoic Acid
CAS 50-82-8
Molekylær formel C7H3O2Cl3
Mængde 50 mg
InChI formel InChI=1S/C7H3Cl3O2/c8-4-2-6(10)5(9)1-3(4)7(11)12/h1-2H,(H,11,12)
SMIL C1=C(C(=CC(=C1Cl)Cl)Cl)C(=O)O
IUPAC navn 2,4,5-trichlorobenzoic acid
Molekylvægt (g/mol) 225.45
Formel vægt 223.9199
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.