missing translation for 'onlineSavingsMsg'
Få mere at vide

2,4,5-Trichloro-1,3-benzenedicarbonitrile, TRC

Artikelnummer. 30454131
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30454131 10 mg
30451214 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30454131 Leverandør Toronto Research Chemicals Leverandørnr. TRCT77385010MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 23039-03-4
Molekylær formel C8HCl3N2
Molekylvægt (g/mol) 231.47
Synonym 2,4,5-Trichloroisophthalonitrile, 2,5,6-Trichloro-1,3-dicyanobenzene, 6-Dechloro Chlorothalonil
IUPAC navn 2,4,5-trichlorobenzene-1,3-dicarbonitrile
SMIL C1=C(C(=C(C(=C1Cl)Cl)C#N)Cl)C#N

Specifications

Kemisk navn eller materiale 2,4,5-Trichloro-1,3-benzenedicarbonitrile
Mængde 10 mg
InChI formel InChI=1S/C8HCl3N2/c9-6-1-4(2-12)7(10)5(3-13)8(6)11/h1H
Formel vægt 229.9205
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.