Få mere at vide
2,4,5-Trichloro-1,3-benzenedicarbonitrile, TRC
Artikelnummer. 30451214
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30451214 100 mg
30454131 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30451214 Leverandør Toronto Research Chemicals Leverandørnr. TRCT773850100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 23039-03-4
Molekylær formel C8HCl3N2
Molekylvægt (g/mol) 231.47
Synonym 2,4,5-Trichloroisophthalonitrile, 2,5,6-Trichloro-1,3-dicyanobenzene, 6-Dechloro Chlorothalonil
IUPAC navn 2,4,5-trichlorobenzene-1,3-dicarbonitrile
SMIL C1=C(C(=C(C(=C1Cl)Cl)C#N)Cl)C#N
Tekniske data
Kemisk navn eller materiale 2,4,5-Trichloro-1,3-benzenedicarbonitrile
Mængde 100 mg
InChI formel InChI=1S/C8HCl3N2/c9-6-1-4(2-12)7(10)5(3-13)8(6)11/h1H
Formel vægt 229.9205