Få mere at vide
2,4,5,6-Tetrachloro-m-xylene, TRC
Artikelnummer. 30426587
Skift visning
Mængde:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426587 250 mg
30408877 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30426587 Leverandør Toronto Research Chemicals Leverandørnr. TRCT291555250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 877-09-8
Molekylær formel C8 H6 Cl4
Molekylvægt (g/mol) 243.95
Synonym Benzene, 1,2,3,5-tetrachloro-4,6-dimethyl-, m-Xylene, 2,4,5,6-tetrachloro- (7CI,8CI), m-Xylene, tetrachloro- (4CI), 1,2,3,5-Tetrachloro-4,6-dimethylbenzene, 1,2,3,5-Tetrachloroxylene, 2,4,5,6-Tetrachloro-m-xylene, 2,4,5,6-Tetrachloro-m-xylene, 2,4,5,6-Tetrachloroxylene, TCMX, Tetrachloro-m-xylene
IUPAC navn 1,2,3,5-tetrachloro-4,6-dimethylbenzene
SMIL Cc1c(Cl)c(C)c(Cl)c(Cl)c1Cl
Tekniske data
Kemisk navn eller materiale 2,4,5,6-Tetrachloro-m-xylene
Mængde 250 mg
InChI formel InChI=1S/C8H6Cl4/c1-3-5(9)4(2)7(11)8(12)6(3)10/h1-2H3
Formel vægt 241.9224