missing translation for 'onlineSavingsMsg'
Learn More

2,4,4'-Trichlorobiphenyl, TRC

Product Code. 30456399
Change view
Click to view available options
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30456399 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456399 Supplier Toronto Research Chemicals Supplier No. TRCT774035500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale PCB No. 28
CAS 7012-37-5
Renhedsgrad noter HPLC
Molekylær formel C12 H7 Cl3
Mængde 500 mg
Synonym 1,1'-Biphenyl','2,4,4'-trichloro-, Biphenyl','2,4,4'-trichloro- (7CI), 2,4,4'-Trichloro-1,1'-biphenyl, 2,4,4'-PCB, 2,4,4'-Trichlorobiphenyl, 2',4,4'-Trichlorobiphenyl, 4,2',4'-Trichloro-1,1'-biphenyl, 4,2',4'-Trichlorobiphenyl, CB 28, K 28, PCB 28, 2,4,4'-TriCB
InChI formel InChI=1S/C12H7Cl3/c13-9-3-1-8(2-4-9)11-6-5-10(14)7-12(11)15/h1-7H
SMIL Clc1ccc(cc1)c2ccc(Cl)cc2Cl
IUPAC navn 2,4-dichloro-1-(4-chlorophenyl)benzene
Molekylvægt (g/mol) 257.54
Formel vægt 255.9613
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.