Få mere at vide
2-(4-(4-Nitrophenyl)butan-2-ylamino)-1-(4-methoxyphenyl)ethanol Hydrochloride, TRC
Artikelnummer. 30482266
Skift visning
Mængde:
5 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482266 5 mg
30471216 250 mg
30467256 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30482266 Leverandør Toronto Research Chemicals Leverandørnr. TRCN5030705MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346600-92-7
Molekylær formel C19H25ClN2O4
Molekylvægt (g/mol) 380.87
IUPAC navn 1-(4-methoxyphenyl)-2-((4-(4-nitrophenyl)butan-2-yl)amino)ethan-1-ol hydrochloride
SMIL CC(CCC1=CC=C(C=C1)[N+](=O)[O-])NCC(C2=CC=C(C=C2)OC)O.Cl
Tekniske data
Kemisk navn eller materiale 2-(4-(4-Nitrophenyl)butan-2-ylamino)-1-(4-methoxyphenyl)ethanol Hydrochloride
Mængde 5 mg
InChI formel InChI=1S/C19H24N2O4.ClH/c1-14(3-4-15-5-9-17(10-6-15)21(23)24)20-13-19(22)16-7-11-18(25-2)12-8-16;/h5-12,14,19-20,22H,3-4,13H2,1-2H3;1H
Formel vægt 380.15