missing translation for 'onlineSavingsMsg'
Learn More

2,4,4,6-Tetrabromo-2,5-cyclohexadien-1-one, 95%, Thermo Scientific Chemicals

Catalog Number p-195501
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
20244-61-5
C6H2Br4O
409.69
MFCD00001589
NJQJGRGGIUNVAB-UHFFFAOYSA-N
2,4,4,6-tetrabromocyclohexa-2,5-dienone, 2,4,4,6-tetrabromo-2,5-cyclohexadienone, 2,4,4,6-tetrabromo-2,5-cyclohexadien-1-one, 2,5-cyclohexadien-1-one, 2,4,4,6-tetrabromo, 2,2,4,4-tetrabromo-2,5-cyclohexadienone, tbco, acmc-1cce4, ksc496q7n, 2,4,4,6-tetrabromocyclohexadienone, 2,4,4,6-tetrabromocyclohexadiene-1-one
88433
2,4,4,6-tetrabromocyclohexa-2,5-dien-1-one
C1=C(C(=O)C(=CC1(Br)Br)Br)Br
This item is not returnable. View return policy
20244-61-5
C6H2Br4O
409.69
MFCD00001589
NJQJGRGGIUNVAB-UHFFFAOYSA-N
2,4,4,6-tetrabromocyclohexa-2,5-dienone, 2,4,4,6-tetrabromo-2,5-cyclohexadienone, 2,4,4,6-tetrabromo-2,5-cyclohexadien-1-one, 2,5-cyclohexadien-1-one, 2,4,4,6-tetrabromo, 2,2,4,4-tetrabromo-2,5-cyclohexadienone, tbco, acmc-1cce4, ksc496q7n, 2,4,4,6-tetrabromocyclohexadienone, 2,4,4,6-tetrabromocyclohexadiene-1-one
88433
2,4,4,6-tetrabromocyclohexa-2,5-dien-1-one
C1=C(C(=O)C(=CC1(Br)Br)Br)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 120°C to 127°C
Color Colorless
Elemental Analysis 94% min.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Iodometry)
Packaging Glass bottle
Quantity 5 g
Fieser 12,457; 16,316
Formula Weight 409.69
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2,4,4,6-Tetrabromo-2,5-cyclohexadien-1-one, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4,4,6-Tetrabromo-2,5-cyclohexadien-1-one
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'