missing translation for 'onlineSavingsMsg'
Få mere at vide

2-[[4-[[2-(1,1-Dimethylethoxy)phenyl]amino]-6-(1-pyrrolidinyl)-1,3,5-triazin-2-yl]amino]ethanol, TRC

Artikelnummer. 30377251
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30377251 10 mg
30378398 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30377251 Leverandør Toronto Research Chemicals Leverandørnr. TRCD47296510MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 958445-02-8
Molekylær formel C19H28N6O2
Molekylvægt (g/mol) 372.46
Synonym 2-[[4-[2-[(2-Methylpropan-2-yl)oxy]anilino]-6-pyrrolidin-1-yl-,1,3,5-triazin-2-yl]amino]ethanol, CID-17757274
SMIL CC(C)(C)Oc1ccccc1Nc2nc(NCCO)nc(n2)N3CCCC3

Tekniske data

Kemisk navn eller materiale 2-((4-((2-(1,1-Dimethylethoxy)phenyl)amino)-6-(1-pyrrolidinyl)-1,3,5-triazin-2-yl)amino)ethanol
Mængde 10 mg
InChI formel InChI=1S/C19H28N6O2/c1-19(2,3)27-15-9-5-4-8-14(15)21-17-22-16(20-10-13-26)23-18(24-17)25-11-6-7-12-25/h4-5,8-9,26H,6-7,10-13H2,1-3H3,(H2,20,21,22,23,24)
Formel vægt 372.2274
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.