Få mere at vide
2-[4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetamide, Mikromol™
Artikelnummer. 30050701
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050701 100 mg
30049101 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050701 Leverandør Mikromol Leverandørnr. MM0177.07
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-[4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetamide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetamide
CAS 55699-13-3
Analyt- eller komponentnavne 2-[4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetamide
Forsendelsestilstand Room Temperature
Molekylær formel C14 H21 N O2
Certificeringer/compliance ISO 17025
Synonym Oxymetazoline Hydrochloride Imp. C (EP), Oxymetazoline Imp. C (EP), 2-[4-(1,1-Dimethylethyl)-3-hydroxy-2,6-dimethylphenyl]acetamide, Oxymetazoline Hydrochloride Impurity C, Oxymetazoline Impurity C
InChI formel InChI=1S/C14H21NO2/c1-8-6-11(14(3,4)5)13(17)9(2)10(8)7-12(15)16/h6,17H,7H2,1-5H3,(H2,15,16)
SMIL Cc1cc(c(O)c(C)c1CC(=O)N)C(C)(C)C
IUPAC navn 2-(4-tert-butyl-3-hydroxy-2,6-dimethylphenyl)acetamide
Molekylvægt (g/mol) 235.32
Mængde 100 mg
Formel vægt 235.1572 g/mol