Få mere at vide
2-[4-(1,1-Dicarboethoxy)benzyl]-2-methyl Malonic Acid-d3, TRC
Artikelnummer. 30371112
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30371112 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371112 Leverandør Toronto Research Chemicals Leverandørnr. TRCD43098225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[4-(1,1-Dicarboethoxy)benzyl]-2-methyl Malonic Acid-d3
CAS 1346604-53-2
Molekylær formel C15H13D3O8
Mængde 25 mg
Synonym 2-[4-(2,2-Dicarboxy-propyl)-phenyl]-2-methylmalonic Acid-d3, [[4-(1,1-Dicarboxyethyl)phenyl]methyl]methyl-propanedioic Acid-d3
InChI formel InChI=1S/C15H16O8/c1-14(10(16)17,11(18)19)7-8-3-5-9(6-4-8)15(2,12(20)21)13(22)23/h3-6H,7H2,1-2H3,(H,16,17)(H,18,19)(H,20,21)(H,22,23)/i2D3
SMIL [2H]C([2H])([2H])C(C(=O)O)(C(=O)O)c1ccc(CC(C)(C(=O)O)C(=O)O)cc1
IUPAC navn 2-[[4-(1,1-dicarboxy-2,2,2-trideuterioethyl)phenyl]methyl]-2-methylpropanedioic acid
Molekylvægt (g/mol) 327.3
Formel vægt 327.1033