missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Pyridyl)-1H-anthra[1,2-d]imidazole-6,11-dione, 97%, Thermo Scientific™

Catalog Number 15441626
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
942119-32-6
C20H11N3O2
325.327
MFCD20265346
IOBRUMYBBWCOCR-UHFFFAOYSA-N
2-3-pyridyl-1h-anthra 1,2-d imidazole-6,11-dione, 2-pyridin-3-yl-1h-anthra 1,2-d imidazole-6,11-dione
16659185
2-pyridin-3-yl-3H-naphtho[3,2-e]benzimidazole-6,11-dione
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C4=C(C=C3)NC(=N4)C5=CN=CC=C5
This item is not returnable. View return policy
942119-32-6
C20H11N3O2
325.327
MFCD20265346
IOBRUMYBBWCOCR-UHFFFAOYSA-N
2-3-pyridyl-1h-anthra 1,2-d imidazole-6,11-dione, 2-pyridin-3-yl-1h-anthra 1,2-d imidazole-6,11-dione
16659185
2-pyridin-3-yl-3H-naphtho[3,2-e]benzimidazole-6,11-dione
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C4=C(C=C3)NC(=N4)C5=CN=CC=C5

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 325.33
Percent Purity 97%
Chemical Name or Material 2-(3-Pyridyl)-1H-anthra[1,2-d]imidazole-6,11-dione
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'