Få mere at vide
2,3-Pentanedione-1,1,1,4,4-d5, TRC
Artikelnummer. 30417478
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30417478 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30417478 Leverandør Toronto Research Chemicals Leverandørnr. TRCP273422100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,3-Pentanedione-1,1,1,4,4-d5
CAS 352431-46-0
Molekylær formel C5 D5 H3 O2
Mængde 100 mg
Synonym 2,3-Pentanedione-1,1,1,4,4-d5 (9CI)
InChI formel InChI=1S/C5H8O2/c1-3-5(7)4(2)6/h3H2,1-2H3/i2D3,3D2
SMIL [2H]C([2H])([2H])C(=O)C(=O)C([2H])([2H])C
IUPAC navn 1,1,1,4,4-pentadeuteriopentane-2,3-dione
Molekylvægt (g/mol) 105.15
Formel vægt 105.0838