Learn More
2',3'-O-Isopropylideneguanosine, TRC
Specifications
Specifications
| Chemical Name or Material | 2',3'-O-Isopropylideneguanosine |
| CAS | 362-76-5 |
| Molecular Formula | C13 H17 N5 O5 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C13H17N5O5/c1-13(2)22-7-5(3-19)21-11(8(7)23-13)18-4-15-6-9(18)16-12(14)17-10(6)20/h4-5,7-8,11,19H,3H2,1-2H3,(H3,14,16,17,20)/t5-,7-,8-,11-/m1/s1 |
| SMILES | CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)n3cnc4C(=O)NC(=Nc34)N |
| IUPAC Name | 9-[(3aR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl]-2-amino-1H-purin-6-one |
| Molecular Weight (g/mol) | 323.3 |
| Formula Weight | 323.123 |