missing translation for 'onlineSavingsMsg'
Få mere at vide

2,3-O-(1-Methylethylidene)-Beta-L-erythropentopyranosid-4-ulose Methyl Ether, TRC

Artikelnummer. 30465200
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30465200 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30465200 Supplier Toronto Research Chemicals Supplier No. TRCM304195100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2,3-O-(1-Methylethylidene)-beta-L-erythropentopyranosid-4-ulose Methyl Ether
CAS 22596-25-4
Molekylær formel C9H14O5
Mængde 100 mg
Synonym 4H-1,3-Dioxolo[4,5-c]pyran','β-L-erythro-pentopyranosid-4-ulose Deriv, β-2,3-O-Isopropylidene-L-erythro-pentopyranosid-4-ulose
InChI formel InChI=1S/C9H14O5/c1-9(2)13-6-5(10)4-12-8(11-3)7(6)14-9/h6-8H,4H2,1-3H3/t6-,7-,8-/m0/s1
SMIL CO[C@@H]1[C@]2([H])[C@@](C(CO1)=O)([H])OC(C)(O2)C
IUPAC navn (3aS,4S,7aR)-4-methoxy-2,2-dimethyldihydro-4H-[1,3]dioxolo[4,5-c]pyran-7(6H)-one
Molekylvægt (g/mol) 202.2
Formel vægt 202.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.