missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Methoxyphenyl)ethylamine, 97+%, Thermo Scientific Chemicals

Catalog Number 15491729
Click to view available options
Quantity:
5 g
100 g
This item is not returnable. View return policy
Regulatory: The product is controlled by Misuse of Drugs Act 1971 and the Misuse of Drugs Regulations 2001, all customers must complete a declaration before delivery.
2039-67-0
C9H13NO
151.209
MFCD00008187
WJBMRZAHTUFBGE-UHFFFAOYSA-N
3-methoxyphenethylamine, 3-methoxyphenylethylamine, 2-3-methoxyphenyl ethanamine, 2-3-methoxyphenyl ethylamine, m-methoxyphenethylamine, m-methoxyphenylethylamine, 2-3-methoxyphenyl ethan-1-amine, benzeneethanamine, 3-methoxy, phenethylamine, m-methoxy, 2-3-methoxy-phenyl-ethylamine
74866
2-(3-methoxyphenyl)ethanamine
COC1=CC=CC(=C1)CCN
This item is not returnable. View return policy
Regulatory: The product is controlled by Misuse of Drugs Act 1971 and the Misuse of Drugs Regulations 2001, all customers must complete a declaration before delivery.
2039-67-0
C9H13NO
151.209
MFCD00008187
WJBMRZAHTUFBGE-UHFFFAOYSA-N
3-methoxyphenethylamine, 3-methoxyphenylethylamine, 2-3-methoxyphenyl ethanamine, 2-3-methoxyphenyl ethylamine, m-methoxyphenethylamine, m-methoxyphenylethylamine, 2-3-methoxyphenyl ethan-1-amine, benzeneethanamine, 3-methoxy, phenethylamine, m-methoxy, 2-3-methoxy-phenyl-ethylamine
74866
2-(3-methoxyphenyl)ethanamine
COC1=CC=CC(=C1)CCN

Activated phenethylamine used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. Also used in a synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Activated phenethylamine used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. Also used in a synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color Yellow
Density 1.038 g/mL
Boiling Point 118°C to 118°C (6 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.538
Quantity 5 g
UN Number UN2735
Beilstein 775202
Solubility Information Insoluble in water.
Formula Weight 151.21
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 2-(3-Methoxyphenyl)ethylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'