missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Methoxyphenyl)ethylamine, ≥97%

Brand:  Alfa Aesar™ H64494.14

Additional Details : CAS Number : 2039-67-0 Weight : 0.02500kg Transport : UN number : 2735 Chem class : 8 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15401739

  • 827.65 DKK / 25g


In Stock 2
Add to basket

Description

Description

Applications
Activated phenethylamine used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. Also used in a synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

2039-67-0
151.209
WJBMRZAHTUFBGE-UHFFFAOYSA-N
74866
COC1=CC=CC(=C1)CCN
C9H13NO
MFCD00008187
3-methoxyphenethylamine, 3-methoxyphenylethylamine, 2-3-methoxyphenyl ethanamine, 2-3-methoxyphenyl ethylamine, m-methoxyphenethylamine, m-methoxyphenylethylamine, 2-3-methoxyphenyl ethan-1-amine, benzeneethanamine, 3-methoxy, phenethylamine, m-methoxy, 2-3-methoxy-phenyl-ethylamine
2-(3-methoxyphenyl)ethanamine

Specifications

2-(3-Methoxyphenyl)ethylamine
1.5380
Yellow
2039-67-0
MFCD00008187
775202
WJBMRZAHTUFBGE-UHFFFAOYSA-N
2-(3-methoxyphenyl)ethanamine
74866
Liquid
1.038g/mL
118°C to 118°C (6mmHg)
25g
C9H13NO
UN2735
3-methoxyphenethylamine, 3-methoxyphenylethylamine, 2-3-methoxyphenyl ethanamine, 2-3-methoxyphenyl ethylamine, m-methoxyphenethylamine, m-methoxyphenylethylamine, 2-3-methoxyphenyl ethan-1-amine, benzeneethanamine, 3-methoxy, phenethylamine, m-methoxy, 2-3-methoxy-phenyl-ethylamine
COC1=CC=CC(=C1)CCN
151.209
151.21
≥97%
Videos
SDS