Få mere at vide
[2-(3-Methoxyphenyl)cyclohex-1-enyl]-N,N-dimethylmethanamine Hydrochloride, Mikromol™
Beskrivelse
[2-(3-Methoxyphenyl)cyclohex-1-enyl]-N,N-dimethylmethanamine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (2-(3-Methoxyphenyl)cyclohex-1-enyl)-N,N-dimethylmethanamine Hydrochloride |
| CAS | 66170-32-9 |
| Analyt- eller komponentnavne | (2-(3-Methoxyphenyl)cyclohex-1-enyl)-N,N-dimethylmethanamine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H23 N O . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Tramadol Hydrochloride Imp. B (EP), Tramadol Imp. B (EP), [2-(3-Methoxyphenyl)cyclohex-1-enyl]-N,N-dimethylmethanamine Hydrochloride, Tramadol Hydrochloride Imp. B (EP) as Hydrochloride, Tramadol Hydrochloride Impurity B as Hydrochloride, Tramadol Impurity B as Hydrochloride |
| InChI formel | InChI=1S/C16H23NO.ClH/c1-17(2)12-14-7-4-5-10-16(14)13-8-6-9-15(11-13)18-3;/h6,8-9,11H,4-5,7,10,12H2,1-3H3;1H |
| SMIL | Cl.COc1cccc(c1)C2=C(CN(C)C)CCCC2 |
| IUPAC navn | 1-[2-(3-methoxyphenyl)cyclohexen-1-yl]-N,N-dimethylmethanamine;hydrochloride |
| Vis mere |