Få mere at vide
[2-(3-Hydroxyphenyl)cyclohex-1-enyl]-N,N-dimethylmethanamine Hydrochloride, Mikromol™
Artikelnummer. 30051053
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051053 100 mg
30049573 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051053 Leverandør Mikromol Leverandørnr. MM0007.50
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
[2-(3-Hydroxyphenyl)cyclohex-1-enyl]-N,N-dimethylmethanamine Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2-(3-Hydroxyphenyl)cyclohex-1-enyl)-N,N-dimethylmethanamine Hydrochloride
CAS 16160-12-6
Analyt- eller komponentnavne (2-(3-Hydroxyphenyl)cyclohex-1-enyl)-N,N-dimethylmethanamine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C15 H21 N O . Cl H
Certificeringer/compliance ISO 9001
Synonym [2-(3-Hydroxyphenyl)cyclohex-1-enyl]-N,N-dimethylmethanamine Hydrochloride
InChI formel InChI=1S/C15H21NO.ClH/c1-16(2)11-13-6-3-4-9-15(13)12-7-5-8-14(17)10-12;/h5,7-8,10,17H,3-4,6,9,11H2,1-2H3;1H
SMIL Cl.CN(C)CC1=C(CCCC1)c2cccc(O)c2
IUPAC navn 3-[2-[(dimethylamino)methyl]cyclohexen-1-yl]phenol;hydrochloride
Molekylvægt (g/mol) 267.79
Mængde 100 mg
Formel vægt 267.139 g/mol