Få mere at vide
2-(3-Ethoxy-4-methoxyphenyl)ethyl-1,1,2,2-d4-amine HCl, CDN
Artikelnummer. 30324212
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30324212 0.1 g 0.1g
30315016 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324212 Leverandør CDN Leverandørnr. CDND73760.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(3-Ethoxy-4-methoxyphenyl)ethanamine-D4 (aminoethyl-D4) Hydrochloride
CAS 1398066-01-7
Molekylær formel C11 D4 H13 N O2 . Cl H
Mængde 0.1 g
Synonym Benzeneethan-α,α,β,β-d4-amine, 3-ethoxy-4-methoxy-, hydrochloride (1:1) (ACI), 3-Ethoxy-4-methoxybenzeneethan-α,α,β,β-d4-amine hydrochloride, α,α,β,β-Tetradeuterio-3-ethoxy-4-methoxyphenylethylamine hydrochloride, 3-O-Ethyl-4-O-methyldopamine-d4 hydrochloride, 3-Ethoxy-4-methoxy-β-phenylethylamine-α,α,β,β-d4 hydrochloride, 2-(3-Ethoxy-4-methoxyphenyl)ethanamine-D4 (aminoethyl-D4) hydrochloride
InChI formel InChI=1 S/C11H17NO2.ClH/c1-3-14-11-8-9(6-7-12)4-5-10(11)13-2;/h4-5,8 H,3,6-7,12H2,1-2H3;1 H/i6D2,7D2;
SMIL Cl.[2 H]C([2 H])(N)C([2 H])([2 H])c1ccc(OC)c(OCC)c1
IUPAC navn 1,1,2,2-tetradeuterio-2-(3-ethoxy-4-methoxyphenyl)ethanamine;hydrochloride
Molekylvægt (g/mol) 235.7438
Formel vægt 235.1277 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity