missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethylphenylacetic acid, 95%, Thermo Scientific Chemicals

Catalog Number 15440939
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
30981-98-7
C10H12O2
164.20
MFCD03844736
AMPNBNGNPLXMOP-UHFFFAOYSA-N
2,3-dimethylphenylacetic acid, 2-2,3-dimethylphenyl acetic acid, 2,3-dimethylbenzeneacetic acid, dimethylbenzeneacetic acid, 2,3-dimethylphenyl acetic acid, 2,3-dimethylphenylaceticacid, xylene-acetic acid, pubchem2466, acmc-1cldr, 2-2,3-dimethylphenyl aceticacid
20572731
2-(2,3-dimethylphenyl)acetic acid
CC1=C(C)C(CC(O)=O)=CC=C1
This item is not returnable. View return policy
30981-98-7
C10H12O2
164.20
MFCD03844736
AMPNBNGNPLXMOP-UHFFFAOYSA-N
2,3-dimethylphenylacetic acid, 2-2,3-dimethylphenyl acetic acid, 2,3-dimethylbenzeneacetic acid, dimethylbenzeneacetic acid, 2,3-dimethylphenyl acetic acid, 2,3-dimethylphenylaceticacid, xylene-acetic acid, pubchem2466, acmc-1cldr, 2-2,3-dimethylphenyl aceticacid
20572731
2-(2,3-dimethylphenyl)acetic acid
CC1=C(C)C(CC(O)=O)=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Physical Form Powder
Formula Weight 164.2
Percent Purity 95%
Chemical Name or Material 2,3-Dimethylphenylacetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'