missing translation for 'onlineSavingsMsg'
Learn More
2,3-Dimethylpentane, 97% Product Code.: 11490600

2,3-Dimethylpentane, 97%

Product Code. 11490600
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11490600

Brand: Thermo Scientific Alfa Aesar 043707.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 565-59-3 | C7H16 | 100.21 g/mol

It is used in the liquid-phase oxidation of 2,4-dimethylpentane and in a study on the preparation and the physical constants of a number of alkanes and cycloalkanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-59-3
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009337
InChI Key WGECXQBGLLYSFP-UHFFFAOYNA-N
Synonym pentane, 2,3-dimethyl, 3,4-dimethylpentane, acmc-1an9i, 3-01-00-00445 beilstein handbook reference, 2,3-dimethylpentane, 2,3-dimethylpentane, analytical standard
PubChem CID 11260
IUPAC Name 2,3-dimethylpentane
SMILES CCC(C)C(C)C

Specifications

Density 0.690g/mL
Boiling Point 89°C to 90°C
Flash Point −6°C (21°F)
Refractive Index 1.3920
Quantity 1 g
UN Number UN1206
Formula Weight 100.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2,3-Dimethylpentane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Product Code
Lot NumberQ14L051 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code043707.03,043707.14,043707.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3-Dimethylpentane, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.