Learn More
2,3-Dimethylmaleic anhydride, 97%, ACROS Organics™
454.00 DKK - 2994.00 DKK
Chemical Identifiers
CAS 766-39-2
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00005523
InChI Key MFGALGYVFGDXIX-UHFFFAOYSA-N
Synonym 2,3-dimethylmaleic anhydride, dimethylmaleic anhydride, 2,5-furandione, 3,4-dimethyl, pyrocinchonic anhydride, maleic anhydride, dimethyl, dimethylmaleic acid anhydride, 2,3-dimethylmaleicanhydride, unii-6pp3n541qa, 3,4-dimethyl-furan-2,5-dione, alpha,beta-dimethylmaleic anhydride
PubChem CID 13010
IUPAC Name 3,4-dimethylfuran-2,5-dione
SMILES CC1=C(C(=O)OC1=O)C
View More Specs
Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10030860
SDS View Documents
Acros Organics
116390050
5g Glass bottle
454.00 DKK
5g
View alternative products
Save Up to 
»
In stock, ready to ship: 4 Please to purchase this item. Need a web account? Register with us today!
10469661
SDS View Documents
Acros Organics
116390500
50g Glass bottle
2994.00 DKK
50g
View alternative products
Save Up to 
»
Estimated Shipment
22-03-2021
Specifications
Chemical Identifiers
766-39-2
126.11
MFGALGYVFGDXIX-UHFFFAOYSA-N
13010
CC1=C(C(=O)OC1=O)C
C6H6O3
MFCD00005523
2,3-dimethylmaleic anhydride, dimethylmaleic anhydride, 2,5-furandione, 3,4-dimethyl, pyrocinchonic anhydride, maleic anhydride, dimethyl, dimethylmaleic acid anhydride, 2,3-dimethylmaleicanhydride, unii-6pp3n541qa, 3,4-dimethyl-furan-2,5-dione, alpha,beta-dimethylmaleic anhydride
3,4-dimethylfuran-2,5-dione
Specifications
2, 3-Dimethylmaleic anhydride
223.0°C
5g
766-39-2
100.0
C6H6O3
17, 445
Solubility in water: slightly soluble. Other solubilities: freely soluble in ethanol,ether and chloroform
CC1=C(C(=O)OC1=O)C
126.11
126.11
Glass bottle
91.0°C to 96.0°C
97%
96.0
97%
MFCD00005523
2,3-dimethylmaleic anhydride, dimethylmaleic anhydride, 2,5-furandione, 3,4-dimethyl, pyrocinchonic anhydride, maleic anhydride, dimethyl, dimethylmaleic acid anhydride, 2,3-dimethylmaleicanhydride, unii-6pp3n541qa, 3,4-dimethyl-furan-2,5-dione, alpha,beta-dimethylmaleic anhydride
MFGALGYVFGDXIX-UHFFFAOYSA-N
3,4-dimethylfuran-2,5-dione
13010
97%
Safety and Handling
Safety and Handling
Product Identifier
  • 2,3-Dimethylmaleic anhydride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.