Få mere at vide
2,3-Diketogulonic Acid Potassium Salt (Technical Grade), TRC
Artikelnummer. 30008211
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008211 50 mg
30011345 10 mg
30008290 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008211 Leverandør Toronto Research Chemicals Leverandørnr. TRCD46792550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2062656-92-0
Molekylær formel C6 H7 O7 . K
Molekylvægt (g/mol) 230.21
Synonym 2,3-Diketogulonic Acid Potassium Salt (Technical Grade), Potassium (4R,5S)-4,5,6-Trihydroxy-2,3-dioxohexanoate
IUPAC navn potassium;(4R,5S)-4,5,6-trihydroxy-2,3-dioxo-hexanoate
SMIL [K+].OC[C@H](O)[C@@H](O)C(=O)C(=O)C(=O)[O-]
Tekniske data
Kemisk navn eller materiale 2,3-Diketogulonic Acid Potassium Salt
Mængde 50 mg
InChI formel InChI=1S/C6H8O7.K/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h2-3,7-9H,1H2,(H,12,13);/q;+1/p-1/t2-,3+;/m0./s1
Formel vægt 229.9829