missing translation for 'onlineSavingsMsg'
Få mere at vide

2,3-Dihydroxy-6-methyl-7-(phenylmethyl)-4-propyl-1-naphthalenecarboxylic Acid, TRC

Artikelnummer. 30375762
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30375762 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375762 Supplier Toronto Research Chemicals Supplier No. TRCD4532655MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2,3-Dihydroxy-6-methyl-7-(phenylmethyl)-4-propyl-1-naphthalenecarboxylic Acid
CAS 213971-34-7
Molekylær formel C22 H22 O4
Mængde 5 mg
Synonym FX11
InChI formel InChI=1S/C22H22O4/c1-3-7-16-17-10-13(2)15(11-14-8-5-4-6-9-14)12-18(17)19(22(25)26)21(24)20(16)23/h4-6,8-10,12,23-24H,3,7,11H2,1-2H3,(H,25,26)
SMIL CCCc1c(O)c(O)c(C(=O)O)c2cc(Cc3ccccc3)c(C)cc12
IUPAC navn 7-benzyl-2,3-dihydroxy-6-methyl-4-propylnaphthalene-1-carboxylic acid
Molekylvægt (g/mol) 350.41
Formel vægt 350.1518
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.