Få mere at vide
2,3-Dihydroxy-1,4-naphthoquinone, TRC
Artikelnummer. 30367023
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367023 10 mg
30354618 50 mg
30360355 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30367023 Leverandør Toronto Research Chemicals Leverandørnr. TRCD36051010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 605-37-8
Molekylær formel C10H6O4
Molekylvægt (g/mol) 190.15
Synonym 2,3-Dihydroxy-1,4-dihydronaphthalene-1,4-dione, 2,3-Dihydroxy-1,4-naphthoquinone, 2,3-Dihydroxy-1,4-naphthosemiquinone, 2,3-Dihydroxynaphthoquinone, Isonaphthazarin, Isonaphthazarine, NSC 401107
IUPAC navn 2,3-dihydroxynaphthalene-1,4-dione
SMIL OC1=C(O)C(=O)c2ccccc2C1=O
Tekniske data
Kemisk navn eller materiale 2,3-Dihydroxy-1,4-naphthoquinone
Mængde 10 mg
InChI formel InChI=1S/C10H6O4/c11-7-5-3-1-2-4-6(5)8(12)10(14)9(7)13/h1-4,13-14H
Formel vægt 190.0266